STEMdiff™ Forebrain Neuron Maturation Kit
Maturation kit for generation of functional neurons from human ES and iPS cell-derived neuronal precursor cells.
The STEMdiff™ Forebrain Neuron Maturation Kit is used in conjunction with the STEMdiff™ Forebrain Neuron Differentiation Kit to generate a mixed population of forebrain-type (FOXG1-positive) neurons from neural progenitor cells derived from human pluripotent stem cells. The neurons derived are highly pure (≥ 90% class III β-tubulin-positive neurons; <10% GFAP-positive astrocytes), functional and can be maintained long-term in culture. Neurons derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation.
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
- Description:STEMdiff™ forebrain neuron maturation kit
- Size:1 kit