- Description:AggreWell™ EB formation medium
- Size:100 ml
- Environmentally Preferable:
- Cat. No.:MSPP-05893
Defined, serum-free medium for generation and culture of embryoid bodies using AggreWell™ plates.
AggreWell™ EB Formation Medium is a serum-free medium that supports the survival of TeSR™-cultured human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells during generation and subsequent culture of embryoid bodies (EBs).