NeuroCult™ NS-A Proliferation Kit (Human)
Catalog # MSPP-05751
Supplier: Stemcell Technologies
CAS Number:
Specifications
- Description:NeuroCult™ NS-A proliferation kit (Human)
- Size:500 ml
- Cat. No.:MSPP-05751
Specifications
About this item
Medium for expansion of human neural stem and progenitor cells.
NeuroCult™ NS-A Proliferation Kit (Human) is a standardized, serum-free basal medium and supplement for the culture of human neural stem and progenitor cells from normal tissues or tumor samples, in the neurosphere or adherent monolayer system. When supplemented with appropriate cytokines, NeuroCult™ NS-A Proliferation Kit (Human) is optimized to maintain human neural stem cells in culture for extended periods of time without the loss of their self-renewal, proliferation, or differentiation potential.
NOTE: Addition of rh EGF, rh bFGF and heparin is required.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').