Orders LinkContactUsLinkComponent
MethoCult™ H4100 Medium
MethoCult™ H4100 Medium
MethoCult™ H4100, 40 ml
Catalog # MSPP-04100
Marketsource
Supplier #:  04100
MethoCult™ H4100 Medium
Catalog # MSPP-04100
Marketsource
CAS Number:  
MethoCult™ H4100 Medium
MethoCult™ H4100 Medium
MethoCult™ H4100, 40 ml
CAS Number:  
Price
$183.08
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Base methylcellulose medium for human cells.
MethoCult™ H4100 is an incomplete medium that contains 2.6% methylcellulose in Iscove's MDM. MethoCult™ H4100 is suitable for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples, when the appropriate growth factors and supplements are added. This formulation does not contain serum or cytokines and can also be used as a base for cloning cell lines in semi-solid media.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Description:
    MethoCult™ H4100
  • Size:
    40 ml