Orders LinkContactUsLinkComponent
StemSpan™ CC110 Culture Supplement
StemSpan™ CC110 Culture Supplement
StemSpan™ CC110, 1 ml
Catalog # MSPP-02697
Marketsource
Supplier #:  02697
StemSpan™ CC110 Culture Supplement
Catalog # MSPP-02697
Marketsource
CAS Number:  
StemSpan™ CC110 Culture Supplement
StemSpan™ CC110 Culture Supplement
StemSpan™ CC110, 1 ml
CAS Number:  
Price
$1,683.08
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Serum-free culture supplement for expansion of human hematopoietic cells.
StemSpan™ CC110 contains a combination of early-acting recombinant human (rh) cytokines formulated to support the proliferation of human hematopoietic cells. It is supplied as a 100X concentrate. When added to serum-free medium, StemSpan™ CC110 promotes the expansion of CD34+ cells isolated from human cord blood and bone marrow. StemSpan™ CC110 stimulates similar expansion of CD34+ cells compared to StemSpan™ CC100, but with higher purity and is meant for use in short-term cultures to activate stem cell and immature progenitor cycling, without necessarily promoting the proliferation and differentiation of later progenitors.
We recommend using StemSpan™ CC110 in combination with any of the following StemSpan™ media:
StemSpan™ SFEM
StemSpan™ SFEM II
StemSpan™-XF
StemSpan™-AOF
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Description:
    StemSpan™ CC110
  • Size:
    1 ml