Orders LinkContactUsLinkComponent
ImmunoCult™ Dendritic Cell Maturation Supplement
ImmunoCult™ Dendritic Cell Maturation Supplement
ImmunoCult™ Dendritic cell maturation supplement, 0.5 ml
Catalog # MSPP-10989
Marketsource
Supplier #:  10989
ImmunoCult™ Dendritic Cell Maturation Supplement
Catalog # MSPP-10989
Marketsource
CAS Number:  
ImmunoCult™ Dendritic Cell Maturation Supplement
ImmunoCult™ Dendritic Cell Maturation Supplement
ImmunoCult™ Dendritic cell maturation supplement, 0.5 ml
CAS Number:  
Price
Inquire For Price
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Supplement for the maturation of human monocyte-derived dendritic cells.
Use ImmunoCult™ Dendritic Cell Maturation Supplement for the maturation of immature dendritic cells (DCs) to mature DCs. For your convenience, the medium is available either individually or as part of the ImmunoCult™ Dendritic Cell Culture Kit. The ImmunoCult™ Dendritic Cell Maturation Supplement is formulated for use in combination with ImmunoCult™-ACF Dendritic Cell Medium and ImmunoCult™-ACF Dendritic Cell Differentiation Supplement to support the maturation of immature DCs.
For more information on protocols for maturation of DCs using ImmunoCult™ Dendritic Cell Maturation Supplement, please explorethe Product Information Sheet (PIS).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Description:
    ImmunoCult™ Dendritic cell maturation supplement
  • Size:
    0.5 ml