- Description:STEMdiff™ Branching lung organoid kit
- Size:1 kit
- Cat. No.:MSPP-100-0195
Serum-free media for generation and maturation of branching lung organoids from human ES and iPS cells.
STEMdiff™ Branching Lung Organoid Kit is a serum-free medium system for efficient and reproducible generation of branching lung organoids from human embryonic stem (ES) and induced pluripotent stem (iPS) cells through four stages of differentiation: 1) definitive endoderm, 2) anterior foregut endoderm, 3) lung bud organoid, and 4) branching lung organoids. STEMdiff™ Branching Lung Organoid Kit has been optimized for differentiation of cells maintained in mTeSR™1. Resulting organoids will develop proximal and distal-like branching airway epithelial structures expressing EPCAM, NKX2.1, SOX2, SOX9, MUC1, and P63.
When maintained beyond 28 days in a Matrigel® sandwich, there is an increase in expression levels of markers for more mature lung cells, such as SFTPC, SFTPB, and ABCA3. For extended periods of organoid culture (>28 days in the Matrigel® sandwich), the components required for branching lung organoid maturation are available in STEMdiff™ Branching Lung Organoid Maturation Kit.