ContactUsLinkComponent
StemSpan™ Lymphoid Differentiation Coating Material (100X)
StemSpan™ Lymphoid Differentiation Coating Material (100X)
Catalog # MSPP-09925
CAS Number:  
undefined
StemSpan™ Lymphoid Differentiation Coating Material (100X)
Catalog # MSPP-09925
Supplier Number:  09925
CAS Number:  
Specifications
  • Description:
    StemSpan™ Lymphoid differentiation coating material (100X)
  • Size:
    250 UL
  • Cat. No.:
    MSPP-09925
Specifications
About this item
Coating material for the differentiation of lymphoid cells.
Use StemSpan™ Lymphoid Differentiation Coating Material (100X) to coat plates for the expansion and differentiation of human CD34+ hematopoietic progenitor cells to T cells. For your convenience, this supplement is available either individually or as part of the StemSpan™ T Cell Generation Kit or the STEMdiff™ T Cell Kit. In this protocol, CD34+ cells are first differentiated to CD7+CD5+ lymphoid progenitor cells, using StemSpan™ Lymphoid Progenitor Expansion Supplement (10X), and then further directed to differentiate to DP T cells. StemSpan™ Lymphoid Differentiation Coating Material (100X) is intended for use in combination with StemSpan™ T Cell Progenitor Maturation Supplement (10X), StemSpan™ SFEM II medium, and StemSpan™ Lymphoid Progenitor Expansion Supplement (10X).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').