ContactUsLinkComponent
STEMdiff™ Forebrain Neuron Differentiation Kit
STEMdiff™ Forebrain Neuron Differentiation Kit
Catalog # MSPP-08600
CAS Number:  
undefined
STEMdiff™ Forebrain Neuron Differentiation Kit
Catalog # MSPP-08600
Supplier Number:  8600
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    STEMdiff™ Forebrain neuron differentiation kit
  • Size:
    1 kit
  • Cat. No.:
    MSPP-08600
Specifications
About this item
Differentiation kit for the generation of neuronal precursors from human ES and iPS cell-derived neural progenitor cells.
The STEMdiff™ Forebrain Neuron Differentiation Kit is used in conjunction with the STEMdiff™ Forebrain Neuron Maturation Kit to generate a mixed population of forebrain-type (FOXG1-positive) neurons from neural progenitor cells derived from human pluripotent stem cells. This kit is optimized to work with STEMdiff™ SMADi Neural Induction Kit, which supplies the appropriate neural progenitor cells. Neurons derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').