Orders LinkContactUsLinkComponent
N2 Supplement-B
N2 Supplement-B
N2 supplement-B, 5 ml
Catalog # MSPP-07156
Marketsource
Supplier #:  07156
N2 Supplement-B
Catalog # MSPP-07156
Marketsource
CAS Number:  
N2 Supplement-B
N2 Supplement-B
N2 supplement-B, 5 ml
CAS Number:  
Price
$215.38
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
For neural differentiation of mouse and human ES and iPS cells.
N2 Supplement-B, containing iron-poor human apo-transferrin, was developed for the in vitro differentiation of mouse or human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells. N2 Supplement-B can be used in a wide range of protocols to achieve neural induction of human ES/iPS cells and/or downstream differentiation of neural progenitor cells to different neuronal or glial subtypes. N2 Supplement-B is provided as a 100X stock solution.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Description:
    N2 supplement-B
  • Size:
    5 ml
Specifications