ContactUsLinkComponent
StemSpan™ Erythroid Expansion Supplement (100X)
StemSpan™ Erythroid Expansion Supplement (100X)
Catalog # MSPP-02692
CAS Number:  
undefined
StemSpan™ Erythroid Expansion Supplement (100X)
Catalog # MSPP-02692
Supplier Number:  02692
CAS Number:  
Specifications
  • Description:
    StemSpan™ Erythroid expansion supplement (100X)
  • Size:
    1 ml
  • Cat. No.:
    MSPP-02692
  • Supplier no.:
    02692
Specifications
About this item
Serum-free culture supplement for expansion of human erythroid cells.
StemSpan™ Erythroid Expansion Supplement (100X) contains a combination of recombinant human cytokines formulated to selectively promote the expansion and differentiation of erythroid progenitor cells from CD34+ cells isolated from human cord blood (CB) or bone marrow (BM) samples. When added to serum-free medium, StemSpan™ Erythroid Expansion Supplement typically promotes the production of thousands of erythroid cells per input CD34+ cell in 14-day liquid cultures initiated with CD34+ human CB cells.
StemSpan™ Erythroid Expansion Supplement (100X) is intended for use in combination with any of the following StemSpan™ media:
StemSpan™ SFEM
StemSpan™ SFEM II
StemSpan™-XF
StemSpan™-ACF Erythroid Expansion Medium
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').