Orders LinkContactUsLinkComponent
STEMdiff™ Endothelial Differentiation Kit
STEMdiff™ Endothelial Differentiation Kit
STEMdiff™ endothelial differentiation kit, 1 kit
Catalog # MSPP-08005
Marketsource
Supplier #:  08005
STEMdiff™ Endothelial Differentiation Kit
Catalog # MSPP-08005
Marketsource
CAS Number:  
STEMdiff™ Endothelial Differentiation Kit
STEMdiff™ Endothelial Differentiation Kit
STEMdiff™ endothelial differentiation kit, 1 kit
CAS Number:  
Price
$1,736.92
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Serum- and xeno-free kit for differentiation and expansion of human pluripotent stem cell-derived endothelial cells.
STEMdiff™ endothelial differentiation kit includes attachment substrate, animal component-free (ACF) endothelial induction medium, and endothelial expansion medium. It is optimized for the differentiation of human pluripotent stem cells (hPSCs) maintained in mTeSR™1, mTeSR™ plus , or TeSR™-E8™ on Corning® Matrigel® to endothelial-like cells. The kit is designed to be used immediately after early mesoderm induction with STEMdiff™ mesoderm induction medium, available for purchase separately. CollPlant is the manufacturer of the rhCollagen component of the cell attachment substrate. The Product may be used for research purposes only.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Description:
    STEMdiff™ endothelial differentiation kit
  • Size:
    1 kit