Orders LinkContactUsLinkComponent
BrainPhys™ Without Phenol Red
BrainPhys™ without phenol red, 500 ml
  MSPP-05791
Marketsource
 :  05791
BrainPhys™ Without Phenol Red
  MSPP-05791
Marketsource
 :  
BrainPhys™ Without Phenol Red
BrainPhys™ Without Phenol Red
BrainPhys™ without phenol red, 500 ml
 :  
$229.23
Each
Serum-free neurophysiological basal medium for improved neuronal function.
Promote, rather than inhibit, neuronal activity and maturity in cultured primary or human pluripotent stem cell (hPSC)-derived neurons in a phenol red-free environment. BrainPhys™ serum-free neuronal basal medium is optimized to yield a higher proportion of synaptically active neurons by mimicking the central nervous system extracellular environment. Use BrainPhys™ without phenol red to culture primary neurons, differentiate and mature hPSC-derived neurons, record microelectrode array-based neuronal activity, live-fluorescent image neurons, and transdifferentiate somatic cells to neurons without hormonal signaling. To ensure cell survival in long-term serum-free culture, BrainPhys™ without phenol red must be combined with an appropriate serum-replacement supplement, such as NeuroCult™ SM1 neuronal supplement and/or N2 supplement-A.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
  • Description:
    BrainPhys™ without phenol red
  • Size:
    500 ml