ContactUsLinkComponent
PneumaCult™ Ex Plus Medium
PneumaCult™ Ex Plus Medium
Catalog # MSPP-05040
CAS Number:  
undefined
PneumaCult™ Ex Plus Medium
Catalog # MSPP-05040
Supplier Number:  05040
CAS Number:  
Specifications
  • Description:
    PneumaCult™-Ex Plus medium
  • Size:
    1 kit
  • Cat. No.:
    MSPP-05040
Specifications
About this item
Serum- and BPE-free medium for expansion of primary human airway epithelial cells.
PneumaCult™-Ex plus medium is a defined, serum- and BPE-free cell culture medium that supports more expansion of primary human airway and nasal epithelial cells at each passage, compared to other commercially available expansion media. This medium also supports at least two additional passages of cell expansion with better differentiation potential, defined as the ability to form a pseudostratified mucociliary epithelium at the air-liquid interface (ALI) using PneumaCult™-ALI medium or a cuboidal epithelium using PneumaCult™-ALI-S medium. PneumaCult™-Ex plus and either PneumaCult™-ALI or PneumaCult™-ALI-S constitute a fully integrated BPE-free culture system for in vitro human airway modeling. This robust and defined system is a valuable tool for basic respiratory research, toxicity studies, and drug development.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').