Orders LinkContactUsLinkComponent
MethoCult™ H4435 Enriched Medium
MethoCult™ H4435 enriched, 72 ml
Catalog # MSPP-04445
Marketsource
 :  04445
MethoCult™ H4435 Enriched Medium
Catalog # MSPP-04445
Marketsource
CAS Number:  
MethoCult™ H4435 Enriched Medium
MethoCult™ H4435 Enriched Medium
MethoCult™ H4435 enriched, 72 ml
CAS Number:  
Price
$2,020.00
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Pre-aliquoted methylcellulose-based medium with recombinant cytokines for human cells. 24x3ml vials.
MethoCult™ H4435 enriched (MethoCult™ GF+ H4435) is a complete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples. This medium supports optimal growth of erythroid progenitor cells (BFU-E and CFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and multipotential granulocyte, erythroid, macrophage and megakaryocyte progenitor cells (CFU-GEMM). MethoCult™ H4435 Enriched is recommended for use with CD34+ cells and other purified cell populations, and for CFU assays performed as part of LTC-IC assays.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Description:
    MethoCult™ H4435 enriched
  • Size:
    72 ml
Specifications