ContactUsLinkComponent
ErgoClip Flask Micro Balances Holder
ErgoClip Flask Micro Balances Holder
Catalog # 97044-244
Supplier:  Mettler Toledo
undefined
ErgoClip Flask Micro Balances Holder
Catalog # 97044-244
Supplier:  Mettler Toledo
Supplier Number:  11107879
Specifications
  • Description:
    Tare container holder for volumetric flasks to use on XPE26/56 micro balances
  • Cat. No.:
    97044-244
  • For flask size:
    1 - 10 ml
  • Supplier no.:
    11107879
Specifications
About this item
ErgoClips micro can be easily assembled on XP26 and XP56 Micro Balances allowing one step dosing directly into the tare container.
  • Fast and safe dosing
  • Accurate weighing
  • Weigh directly into tare containers
The ErgoClips safer dosing method substantially reduces contamination risks, enables fast dosing processes and increases productivity.
Say goodbye to weighing paper! Dosing directly into the tare container eliminates transfer errors and saves money by minimizing sample loss.