External Thermal Printer for Vertical Autoclaves
Catalog # 77574-048
Supplier: TUTTNAUER USA CO. LTD.
New Product
Specifications
- Description:Thermal printer for LabSci autoclave
- Environmentally Preferable:
- Cat. No.:77574-048
Specifications
About this item
External Thermal printer for Vertical LabSci autoclaves. Printer is capable of printing barcode labels or sterilizer cycle information.
Prints cycle data on thermal paper or UDI barcode labels.
Ordering information: The thermal paper (77574-052) is not included and must be ordered separately. Supplied with 1 roll of labels (77574-050).