Order Entry
United States
ContactUsLinkComponent
Eisco® Specific Gravity Bottle
Eisco® Specific Gravity Bottle
Catalog # 470345-862
Supplier:  Wards
undefined
Eisco® Specific Gravity Bottle
Catalog # 470345-862
Supplier:  Wards
Supplier Number:  PH0122B

Specifications

  • Capacity:
    50 ml
  • Description:
    Specific gravity bottle
  • Dimensions:
    4" H (with stopper), Ø 2"
  • Environmentally Preferable:
  • Cat. No.:
    470345-862

Specifications

About this item

Flat-bottomed glass bottle with a perforated stopper for accurate specific gravity measurements.

  • Specific gravity bottle made of high-quality borosilicate 3.3 glass
  • Perforated stopper
  • Flat bottom for spill prevention

Specific gravity bottle made of high quality borosilicate glass. 50 ml capacity with flat bottom. Includes perforated stopper. Unadjusted spherical pattern.