SafeCup™ III 11-Panel All-in-One Cup Tests
Catalog # 77574-668
Supplier: Germaine Laboratories
New Product
Specifications
- Description:SafeCup™ III 11-panel all-in-one cup test
- Size:25 cups/kit
- Cat. No.:77574-668
- Sensitivity:See IFU or contact
- Parameters tested:AMP, BAR, BZO, BUP, COC, MTD, MDMA, MET, MOP, OXY, PCP
Specifications
About this item
Rapid qualitative tests for the detection of drugs of abuse in urine.
- A total collection, testing, and transport system
- Easier to use, easier to read
- Quick results in 5 minutes
- Includes 'Click Seal' cap for improved leak resistance
- Store at room temperature
- Adheres to SAMHSA cutoff levels
- Includes gloves, temperature strip, security seals, IFU
All-in-One cup tests for amphetamine, barbiturates, benzodiazpines, buprenorphine, cocaine, methadone, mdma, methamphetamine, morphine, oxycodone, and phencyclidine. 25 cups per case. CLIA waived.
Certifications: CLIA waived for over the counter and point of care use.
Ordering information: Orders received by 12:30 PM CST process same day. If not in stock, product requires 1 to 2 days lead time.