Human Recombinant DLK (GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
New Product
Certificates
About this item
Recombinant Human DLK (1 to 520) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage Conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein Synonyms:MEKK12|ZPK|MUK|ZPKP1|DLK
- Protein/Peptide Name:Map3k12
- Molecular Weight:94 kDa
- Formulation:Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
- Shipping Temperature:It is highly recommended Enzymes be shipped at frozen temperature with dry ice. Shipment at ambient temperature may seriously affect the activity of the ordered products.