Orders LinkContactUsLinkComponent
Tissue Floating Bath
Tissue floating bath
  77435-368
 :  TF1000
Tissue Floating Bath
  77435-368
Tissue Floating Bath
Tissue Floating Bath
Tissue floating bath
$891.83
Each
The precisionary instruments tissue floating bath is a reliable and efficient solution for softening embedded tissue specimens without causing wrinkles or distortions. With a teflon-coated chamber for easy cleaning, a low-profile interior chamber to reduce glare.
  • The tissue floating bath is used after sectioning with a rotary microtome
For use after sectioning paraffin wax.
  • Description:
    Tissue floating bath