To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Sheep peste des petits ruminants kit
- Environmentally Preferable:
- Storage Conditions:The unopened kit is stable for 6 months from the date of manufacture when stored at 2...8 °C, avoid light, the shelf life is 60 days from the date of opening
- Shipping Conditions:Shipment with room temperature
- Cat. No.:77574-022
- Application:Test sheep samples for peste des petits ruminants virus
This kit is developed for serologic test for sheep IgG titer of Anti-Peste Des Petits Ruminants Virus (PPRV) antibody in serum/plasma or purified human antibody (monoclonal and polyclonal) in vitro.
- This kit was tested to bind to convalescent sheep sera (n=9) with sensitivity of 100%, specificity of 100%
This kit is developed for a standard indirect-ELISA format, providing a rapid detection of anti-PPRV sheep IgG in serum. The kit consists of high-bind pre-coated, detachable 96-well plate, Positive control, Negative Control, an HRP-Anti-sheep IgG secondary antibody, TMB and dilution/wash/stop buffer.
Use protocol (four simple steps):
1. Detach wells from plate based on your samples and keep at room temperature. Dilute your samples, positive and negative controls in sample dilution buffer and add them to wells (0.1 ml/well). Incubate at 37 °C for 1 h or room temperature for 1.5 h. Wash wells four times by wash solution (0.3 ml).
2. Dilute the Secondary HRP antibody by wash solution and add to each well (0.1 ml). Incubate at room temperature for 0.5 to 1 h. Avoid light by foil. Wash wells as mentioned.
3. Add TMB to each well (0.1 ml) and incubate at room temperature for 5 to 30 minutes.
4. Stop the substrate reaction by adding stop solution (0.1 ml/well). Absorbance (OD) is calculated as the absorbance at 450 nm. The OD Value reflects the amount of antibody bound.