Order Entry
Puerto Rico
ContactUsLinkComponent
tert-Butyl 4-(bromomethyl)-1H-pyrazole-1-carboxylate 95%
Corrosive
tert-Butyl 4-(bromomethyl)-1H-pyrazole-1-carboxylate 95%
Catalog # 77565-968
Supplier:  Ambeed
New Product
CAS Number:  
Corrosive
tert-Butyl 4-(bromomethyl)-1H-pyrazole-1-carboxylate 95%
Catalog # 77565-968
Supplier:  Ambeed
Supplier Number:  A1122593-250MG
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    250 mg
  • Cat. No.:
    77565-968

Specifications

About this item

 :
ADR:8,Ⅲ
 :261.12 g/mol
UN:3261
 :GHS05
CAS Number:530144-72-0
Formula:C₉H₁₃BrN₂O₂
MDL Number:MFCD18837735