Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Buffer, TAE Solution 10X Concentrate (TRIS-Acetate EDTA Buffer)

Supplier:   RPI
Buffer, TAE Solution 10X Concentrate (TRIS-Acetate EDTA Buffer)
Supplier:  RPI
Certificates
Buffer, TAE Solution 10X Concentrate (TRIS-Acetate EDTA Buffer)
Buffer, TAE Solution 10X Concentrate (TRIS-Acetate EDTA Buffer)

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Buffer, TRIS acetate EDTA 10X solution.

  • 10X solution contains:
  • TRIS-base: 0.4 M
  • Acetic acid: 0.2 M
  • EDTA: 10 mM
  • pH (10X solution (at 25 °C): 8.2 to 8.4

 : Research or further manufacturing use only, not for food or drug use.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 76348-628
  • 76348-630
  • 77510-472
  • Appearance:
  • Clear, colorless solution
  • Clear, colorless solution
  • Clear, colorless solution
  • Storage Temperature:
  • RT
  • RT
  • RT

Specifications

Product Family Options

Product Information

  • Size
    pH
    Concentration
    Availability
    Price
  • 76348-628
    TRIS acetate EDTA solution
    TRIS acetate EDTA solution
    1 L
    8.2 - 8.4
    10X
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $53.75
    $53.75
    Each (1l)
       $53.75

    Supplier Number: T60010-1000.0
    Specifications:
    More
    Specifications
    Cat. No.
    76348-628
    Appearance
    Clear, colorless solution
    Storage Temperature
    RT
  • 76348-630
    TRIS acetate EDTA solution
    TRIS acetate EDTA solution
    4 L
    8.2 - 8.4
    10X
    Log in for availability
    Log in for availability
    Each (4l)
    Each (4l) - $160.14
    $160.14
    Each (4l)
       $160.14

    Supplier Number: T60010-4000.0
    Specifications:
    More
    Specifications
    Cat. No.
    76348-630
    Appearance
    Clear, colorless solution
    Storage Temperature
    RT
  • 77510-472
    TRIS acetate EDTA solution
    TRIS acetate EDTA solution
    10 L
    8.2 - 8.4
    10X
    Log in for availability
    Log in for availability
    Each
    Each - $276.54
    $276.54
    Each
       $276.54

    Supplier Number: T60010-10000.0
    Specifications:
    More
    Specifications
    Cat. No.
    77510-472
    Appearance
    Clear, colorless solution
    Storage Temperature
    RT

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.