Human Recombinant Tau-441 (Avi tag) (from E. coli), Biotinylated
Supplier: Sino Biological
New Product
Certificates
About this item
Recombinant human Tau-441 expressed in E. coli cells with an N-terminal Avi tag and enzymatically biotinylated in vitro prior to the final chromatography purification.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein Synonyms:PPP1R103|FTDP-17|MAPT|TAU|PPND|MAPTL|MTBT1|MTBT2|MSTD|DDPAC
- Protein/Peptide Name:MAPT
- Molecular Weight:64 kDa (45.9 kDa by MW calculation)
- Formulation:Supplied as sterile PBS.
- Shipping Temperature:It is highly recommended Enzymes be shipped at frozen temperature with dry ice. Shipment at ambient temperature may seriously affect the activity of the ordered products.
Specifications
Frequently Bought Together