Human Recombinant Alpha Synuclein Pre-Formed Fibril (Type 1) (from E. coli)
Supplier: Sino Biological
New Product
Certificates
About this item
Recombinant full length tag-free human Alpha-synuclein Pre-Formed Fibril (Type 1) was expressed in E. coli cells.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein Synonyms:PARK1|PARK4|NACP|PD1
- Protein/Peptide Name:SNCA
- Molecular Weight:14.5 kDa
- Formulation:Supplied as sterile PBS.
- Shipping Temperature:It is highly recommended Enzymes be shipped at frozen temperature with dry ice. Shipment at ambient temperature may seriously affect the activity of the ordered products.