Specifications
- Description:Rapid Response™ Fentanyl test cassette
- Cat. No.:77548-848
Specifications
About this item
The Rapid Response™ Fentanyl Test Cassette is a competitive binding, lateral flow immunochromatographic assay for qualitative detection of fentanyl in human urine at the cut-off concentration of 1 ng/ml. This single drug test cassette can provide accurate and fast results in 5 minutes.
- Sample: Urine
- Format: Cassette
- Rapid test for detecting Fentanyl in human urine
- Read results in 5 minutes
- Individually packaged test cassettes with pipette droppers
To perform the test, place the test cassette on a clean and level surface. Hold the dropper vertically and add 3 drops of urine to the specimen well (S) of the test cassette. Read results at 5 minutes. Interpretation of results are: 2 colored lines, one in Control Region (C) and another in Test Region (T), is negative, 1 line in the Control Region (C) and no line in the Test Region (T) is positive, no lines or no line in the Control Region (C) is invalid.