$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Methanol : Sulfuric acid
Methanol : Sulfuric acid solution, Supelco®
Methanol : Sulfuric acid solution, Supelco®
Methanol : Sulfuric acid solution, Supelco®
Catalog # EM-GC0074-10
Supplier:
MilliporeSigma
CAS Number:
SDS
Certificates
Expand image
Methanol : Sulfuric acid solution, Supelco®
Catalog # EM-GC0074-10
Supplier:
MilliporeSigma
Supplier Number:
GC0074-10
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic barrel
Size:
200 L
Cat. No.:
EM-GC0074-10
Supplier no.:
GC0074-10
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation