Caps for 5-Layer Cell Culture Flasks
Made of high clarity, 100% virgin polystyrene.
- Sterilized by E-beam, SAL= 10⁻⁶
- Non-pyrogenic, DNase/Rnase free
- Growth area: 870 cm²
Vacuum plasma tissue culture treated (TC treated). Individually packaged in sterile bag.