Orders LinkContactUsLinkComponent
NestShield® Extra Thick Nitrile Examination Gloves
NestShield® Extra Thick Nitrile Examination Gloves
NestShield® Extra Thick Nitrile Examination Gloves
NestShield extra thick nitrile examination gloves offer even more protection in a high-quality disposable glove, designed for a great fit no matter how many times you apply or remove them.
  • Fingertip pitting design provides enhanced anti-slip performance
  • A comfortable feel similar to natural latex
  • Tensile Strength: Minimum 14.0 MPa (Before aging)
  • Palm thickness: Minimum 0.09 mm
  • Ambidextrous, standard beaded cuff and fingertip, flexible operation
At 6 mil, these powder-free black gloves give you high-performing nitrile material with extra thickness for heavy-duty performance, whether you are handling heavy machinery or industrial cleaning chemicals.
They are latex-free and powder-free nitrile. That way, you never need to worry about allergic skin reactions.
 : These nitrile gloves have been certified with the FDA 510K registration and meet the rigorous testing standards of the American Society for Testing and Materials.
  • Catalog No:
  • 77508-964
  • 77508-966
  • 77508-968
  • 77508-970
  • 77508-972
  • Ambidextrous/Hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable/Reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex/Latex-free:
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Powdered/Powder-free:
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Cuff Style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
Product Family Options
  • Description
    Glove size
    Glove color
    Length
  • NestShield® Extra thick nitrile examination gloves
    X-Small
    Black
    240 mm
    Case of 1,000
    Case of 1,000 - $160.13
    $160.13
    Case of 1,000
       $160.13
     : 902801
    Specifications
    Cat. No.
    77508-964
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable/Reusable
    Disposable
    Latex/Latex-free
    Latex-Free
    Powdered/Powder-free
    Powder-Free
    Cuff Style
    Beaded
  • NestShield® Extra thick nitrile examination gloves
    Small
    Black
    240 mm
    Case of 1,000
    Case of 1,000 - $160.13
    $160.13
    Case of 1,000
       $160.13
     : 902811
    Specifications
    Cat. No.
    77508-966
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable/Reusable
    Disposable
    Latex/Latex-free
    Latex-Free
    Powdered/Powder-free
    Powder-Free
    Cuff Style
    Beaded
  • NestShield® Extra thick nitrile examination gloves
    Medium
    Black
    240 mm
    Case of 1,000
    Case of 1,000 - $160.13
    $160.13
    Case of 1,000
       $160.13
     : 902821
    Specifications
    Cat. No.
    77508-968
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable/Reusable
    Disposable
    Latex/Latex-free
    Latex-Free
    Powdered/Powder-free
    Powder-Free
    Cuff Style
    Beaded
  • NestShield® Extra thick nitrile examination gloves
    Large
    Black
    240 mm
    Case of 1,000
    Case of 1,000 - $160.13
    $160.13
    Case of 1,000
       $160.13
     : 902831
    Specifications
    Cat. No.
    77508-970
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable/Reusable
    Disposable
    Latex/Latex-free
    Latex-Free
    Powdered/Powder-free
    Powder-Free
    Cuff Style
    Beaded
  • NestShield® Extra thick nitrile examination gloves
    X-Large
    Black
    240 mm
    Case of 900
    Case of 900 - $160.13
    $160.13
    Case of 900
       $160.13
     : 902841
    Specifications
    Cat. No.
    77508-972
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable/Reusable
    Disposable
    Latex/Latex-free
    Latex-Free
    Powdered/Powder-free
    Powder-Free
    Cuff Style
    Beaded