BioFactory™ Systems are compact, multi-layer, single-use cell culture systems designed for easy scale-up cell culture applications such as production of vaccines, monoclonal antibodies or pharmaceuticals. They're versatile and easy-to-use system for medium to large scale research or commercial production with reduced contamination risk. Narrow mouth caps available for tubing solutions.
- Made of high clarity, 100% virgin polystyrene
- Sterilized by E-beam, SAL = 10⁻⁶
- Non-pyrogenic, DNase/Rnase free
- TC treated, good for cell attachment and growth
- Clear lot number for batch traceability
- Ultrasonic welding without extrinsic ingredients
- Excellent welding line design can avoid producing particles during welding and make the biofactory stronger
- Vent caps with 0.22 μm hydrophobic gas permeable membrane, hindering bacteria and water, can also avoid liquid swelling
- Wide mouth is applicable to pour culture medium directly. Narrow mouth is applicable to operate with the aseptic pipeline
- Large growing area
- Applicable to automatic machine
- Standardize operations to reduce batch differences
Growth kinetics are the same as cells grow in the cell culture flasks. Applicable to large-scalely culture adherent cells. Large mouth design for easy pouring of the medium directly and the small mouth design for easy connection with the feeding systems. Solid overcaps can be used directly on the adaptor caps to block dust and bacteria.
BioFactory™ Culture chambers with tube:
Chambers (with tube) realizes the closed transfer of liquid, which can directly transfer the culture medium or cells to a cell factory or a bioreactor through a connecting tube in a sterile environment, greatly reducing the risk of exogenous pollution in the drug development and production process, simultaneously omitting the operations of tube design, assembling and sterilization, therefore, improving the production efficiency.
BioFactory™ Culture chambers with tube features:
Pre-assembled radiation-resistant high throughput air filter, pervious and resistant to bacteria, ensuring sterility and high flow rate.
Closed liquid transfer, avoiding open operation and reducing the risk of pollution in the process of liquid transfer.
The liquid inlet tube can be aseptically welded under a normal environment.
High-quality materials and smooth inner wall of the tube, ensuring an excellent transmission performance.