ContactUsLinkComponent
HiTrap™ Capto™ Phenyl Hydrophobic Interaction Chromatography Columns (High Sub)
HiTrap™ Capto™ Phenyl Hydrophobic Interaction Chromatography Columns (High Sub)
Supplier:  Cytiva
Certificates
undefined
HiTrap™ Capto™ Phenyl Hydrophobic Interaction Chromatography Columns (High Sub)
Supplier:  Cytiva
Specifications
  • Catalog No:
  • 77509-270
  • 77509-272
  • Storage:
  • 4...30 °C, 20% ethanol
  • 4...30 °C, 20% ethanol
Specifications