SP Wilmad-LabGlass Soxhlet Extraction Apparatus, SP Industries
Supplier: SP Industries
Certificates
About this item
These borosilicate glass Soxhlet extraction apparatus are used for the continuous extraction of solids with a suitable solvent
Complete units are supplied with a flat bottom boiling flask (LG-7230-102, LG-7230-104, or LG-7230-108), a Soxhlet extraction tube and an Allihn condenser. The extractor capacity is the solvent volume held in the extractor to the top of the siphon tube. The siphon tube is protected from accidental damage by the vapor tube. The extractor has a standard taper outer joint at the top for connection to the condenser and a standard taper inner joint at the bottom for connection to the flask.
Ordering information: Soxhlet extraction apparatus uses cellulose, glass microfiber or fritted glass extraction thimbles (not supplied).