Accessories for Adjustable Slide Truck, Metro International
Catalog # 19766-010
Supplier: Metro
Specifications
- Depth:29½"
- Width:42½"
- Description:Grounding Bracket and Cable
- Height:38"
- Environmentally Preferable:
- Cat. No.:19766-010
- Shipping weight:0.5 lbs.
- Supplier no.:GCB16S