Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

APS/TEMED

Flammable
Corrosive
Toxic
Supplier:   RPI
APS/TEMED
Supplier:  RPI
Certificates
APS/TEMED

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Tertiary amine used to catalyze formation of free radicals from ammonium persulfate and riboflavin. Suitable for electrophoresis and tested as a catalyst for polyacrylamide gel electrophoresis.

 : Research or further manufacturing use only, not for food or drug use.

Synonyms:  Ammonium persulphate/N,N,N′,N′-Tetramethylethylenediamine, Ammonium peroxodisulphate/N,N,N′,N′-Tetramethylethylenediamine
  • CAS Number:7727-54-0/110-18-9
  • Boiling Pt:120...122 °C
  • Melting Pt:–55 °C
  • Flash Pt:20 °C
  • Storage Temperature:Room Temperature
  • EINECS:203-744-6
  • ADR:3,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear, Colorless Solution
  • Identification:
    Conforms
  • Purity (GC):
    ≥ 97 %
  • BSE/TSE Free:
    Conform
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 25 ml
    Log in for availability
    Log in for availability
    Each
    Each - $34.16
    $34.16
    Each
       $34.16

    Supplier Number: T18000-0.025
  • 50 ml
    Log in for availability
    Log in for availability
    Each
    Each - $53.26
    $53.26
    Each
       $53.26

    Supplier Number: T18000-0.05
  • 100 ml
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $107.75
    $107.75
    Each (100ml)
       $107.75

    Supplier Number: T18000-0.1

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.