Erlenmeyer Flasks, Closed System
Supplier: NEST SCIENTIFIC
Jump to:
About this item
For aseptic transfer of culture medium and cell culture solution.
- Filter membrane area (0.22 μm): Ø 42 mm
- Tube material: TPE
One end of the pipeline is connected to the MPC series connector, which increases the convenience and safety in use. The male connector and the sealing cap are tightly connected to prevent accidental disconnection, and the connector can be rotated to reduce pipeline distortion.
2 Options Available Below
Product Details & Documents
For aseptic transfer of culture medium and cell culture solution.
- Filter membrane area (0.22 μm): Ø 42 mm
- Tube material: TPE
One end of the pipeline is connected to the MPC series connector, which increases the convenience and safety in use. The male connector and the sealing cap are tightly connected to prevent accidental disconnection, and the connector can be rotated to reduce pipeline distortion.
Product Family Options
Product Information
- CapacitySterilityNeck I.D.Neck O.D.Tubing lengthAvailabilityPrice
- 77509-104Erlenmeyer flask with Bi-directional transfer cap, TPE tube, heat sealErlenmeyer flask with Bi-directional transfer cap, TPE tube, heat seal250 mlSterile¹/₈"¹/₄"50 cmLog in for availabilityCase of 4$486.40Supplier Number: C30921-AZC050A
- 77509-114Erlenmeyer flask with Bi-directional transfer cap, TPE tube, heat sealErlenmeyer flask with Bi-directional transfer cap, TPE tube, heat seal1000 mlSterile¹/₈"¹/₄"50 cmLog in for availabilityCase of 4$517.32Supplier Number: C31121-AZC050A