Orders LinkContactUsLinkComponent
Bioact® Cleaner, MicroCare
Flux Remover in Metal Pail, 18.9 L (5 gal.)
Catalog # 46620-666
Supplier:   MicroCare
 :  MCC-EC7MP
Bioact® Cleaner, MicroCare
Catalog # 46620-666
Supplier:  MicroCare
Bioact® Cleaner, MicroCare
Flux Remover in Metal Pail, 18.9 L (5 gal.)
Price
$2,791.93
Log in for availability
Each (5US Gal)
Product Details & Documents
Based on biodegradeable citrus terpenes derived from Florida oranges, this solvent aggressively cleans water-soluble fluxes and pastes.
  • Nontoxic
  • Combustible
  • Ozone-safe
Very effective on rosin, synthetic, and no-clean fluxes. Bioact® replaces traditional CFC solvents, does not need a water rinse, and has a pleasant citrus scent.
Documents
Specifications
  • Description:
    Flux Remover in Metal Pail
  • Size:
    18.9 L (5 gal.)