ContactUsLinkComponent
Konform® AR Conformal Coating, ITW Chemtronics®
Konform® AR Conformal Coating, ITW Chemtronics®
Catalog # 47732-652
Supplier:  ITW Chemtronics
UL Listed
undefined
Konform® AR Conformal Coating, ITW Chemtronics®
Catalog # 47732-652
Supplier:  ITW Chemtronics
Supplier Number:  CTAR-12
UL Listed
Specifications
  • Volume:
    326 g (11.5 oz.)
  • Description:
    Aerosol Can
  • Cat. No.:
    47732-652
  • Supplier no.:
    CTAR-12
Specifications
About this item
Konform® AR is ideal for insulating against high-voltage arcing and corona shorts
UL listed.
This acrylic conformal coating provides a hard, durable protective barrier against humidity, salt, corrosive vapors, and fungus for printed circuit boards and electronic assemblies. Protects and increases life of electronic assemblies. Meets MIL-I-46058C, Type AR. Usable temperature range: –59° to 132°C (–75° to 270°F).