Sample Loop for UW Type Valves
Stainless steel, 250 µl, ¹/₁₆"
Catalog # AGVLSL250CUW
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Supplier #: VLSL250CUW
Jump to:
Product Details & Documents
Stainless steel sample loops for UW type valves.
- Two sets of nuts and ferrules included
Specifications
- Ø int.:¹/₁₆"
- Capacity:250 µl
- Material:Stainless steel