Envi-Ro-Tech™ 1671 Dusters, Techspray™
Supplier: ITW Chemtronics
Certificates
About this item
Exceptionally pure, moisture-free inert gas removes particulates without using harmful solvents.
- Nonabrasive
- Contain no CFCs
Nonflammable dusters leave no residue and are quad-filtered.