ContactUsLinkComponent
Ultrajet® 70 Duster, ITW Chemtronics®
Ultrajet® 70 Duster, ITW Chemtronics®
Catalog # 15145-520
Supplier:  ITW Chemtronics
undefined
Ultrajet® 70 Duster, ITW Chemtronics®
Catalog # 15145-520
Supplier:  ITW Chemtronics
Supplier Number:  ES1015
Specifications
  • Volume:
    284 g (10 oz.)
  • Cat. No.:
    15145-520
  • Supplier no.:
    ES1015
Specifications
About this item
This extra-strength duster removes dust, dirt, and other dry contaminants
Economizes solvent usage by removing layers of dry particulate first so the solvent can work immediately on more difficult-to-remove soils. Ideal for use on keyboards, circuit boards, surface mount devices, computers and other electronic components, assemblies, and photographic and video equipment. Nonflammable. Ozone-safe, CFC- and HCFC-free. 100% ultra-pure HFC-134a leaves no residues. Completely safe for use on most plastics.