Orders LinkContactUsLinkComponent
Ultrajet® Big Blast Dusters, ITW Chemtronics®
Ultrajet® Big Blast Dusters, ITW Chemtronics®
Supplier:   ITW Chemtronics
Ultrajet® Big Blast Dusters, ITW Chemtronics®
Supplier:  ITW Chemtronics
Certificates
Ultrajet® Big Blast Dusters, ITW Chemtronics®
Ultrajet® Big Blast Dusters, ITW Chemtronics®
3 Options Available Below
Product Details & Documents
Dusters deep cleans hard-to-reach areas, removing dirt, dust, and dry contaminants
Complete system includes chrome trigger valve, 91.4cm (36") extension hose, and 284g (10oz.) can. Trigger valve and extension hose fit refill can 15145-500 for economical reuse.
Ideal for use on keyboards, circuit boards, surface mount devices, computers and other electronic components, assemblies, and photographic and video equipment. Nonflammable. Ozone-safe, CFC- and HCFC-free, and nonabrasive. 100% ultra-pure HFC-134a leaves no residues. Completely safe for use on energized equipment and most plastics.
Product Family Options
Product Information
  • Volume
    Availability
    Price
  • 15145-496
    Ultrajet® Complete System
    Ultrajet® Complete System
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $215.04
    $215.04
    Log in for availability
    Case of 12
    Case of 12 - $1,609.73
    $1,609.73
    Each
       $215.04
    Case of 12
       $1,609.73
    Supplier Number: ES1020K
  • 15145-422
    Ultrajet® Duster, Aerosol Can
    Ultrajet® Duster, Aerosol Can
    284 g (10 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $105.83
    $105.83
    Log in for availability
    Case of 12 (1/ Itm/Bx)
    Case of 12 (1/ Itm/Bx) - $623.82
    $623.82
    Each(1/Itm/Bx)
       $105.83
    Case of 12 (1/ Itm/Bx)
       $623.82
    Supplier Number: ES1020
  • 15145-500
    Ultrajet®, Refill Aerosol Can
    Ultrajet®, Refill Aerosol Can
    284 g (10 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $85.09
    $85.09
    Log in for availability
    Case of 12 (1/ Itm/Bx)
    Case of 12 (1/ Itm/Bx) - $520.96
    $520.96
    Each(1/Itm/Bx)
       $85.09
    Case of 12 (1/ Itm/Bx)
       $520.96
    Supplier Number: ES1020R