ContactUsLinkComponent
Hypersil™ PREP BDS C18 HPLC Columns
Hypersil™ PREP BDS C18 HPLC Columns
Supplier:  Thermo Scientific
Certificates
Hypersil™ PREP BDS C18 HPLC Columns
Supplier:  Thermo Scientific
Specifications
  • Catalog No:
  • 76330-142
  • 76330-624
  • 76330-140
  • 76330-144
  • 76330-978
  • 76330-980
  • 76330-628
  • 76330-626
  • 76330-976
  • 76330-156
  • 76330-640
  • 76330-638
  • 76330-154
  • 76330-160
  • 76330-644
  • 76330-642
  • 76330-158
  • 76330-148
  • 76330-630
  • 76330-146
  • 76330-636
  • 76330-982
  • 76330-152
  • 76330-634
  • 76330-150
  • 76330-604
  • 76330-602
  • 76330-606
  • 76330-618
  • 76330-134
  • 76330-132
  • 76330-616
  • 76330-138
  • 76330-622
  • 76330-534
  • 76330-620
  • 76330-136
  • 76330-610
  • 76330-608
  • 76330-614
  • 76330-612
  • 76330-596
  • 76330-594
  • 76330-600
  • 76330-598
  • 76330-592
  • 76330-590
  • 76330-164
  • 76330-162
  • 76330-646
  • 76330-166
  • 76330-650
  • 76330-648
  • 76330-662
  • 76330-660
  • 76330-664
  • 76330-170
  • 76330-654
  • 76330-652
  • 76330-168
  • 76330-658
  • 76330-656
  • Description:
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, XtendedLife
  • Preparative column, XtendedLife
  • Guard
  • Preparative column, standard
  • Preparative column, XtendedLife
  • Preparative column, standard
  • Preparative column, XtendedLife
  • Preparative column, XtendedLife
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, XtendedLife
  • Preparative column, XtendedLife
  • Preparative column, standard
  • Preparative column, standard
  • Guard
  • Preparative column, standard
  • Guard ¹/₁₆"
  • Preparative column, XtendedLife
  • Preparative column, standard
  • Guard ¹/₁₆"
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, standard
  • Preparative column, XtendedLife
  • Preparative column, standard
  • Preparative column, XtendedLife
  • Preparative column, XtendedLife
  • Guard ¹/₈"
  • Preparative column, XtendedLife
  • Guard ¹/₈"
  • Preparative column, standard
  • Preparative column, XtendedLife
  • Preparative column, XtendedLife
  • Preparative column, standard
  • Preparative column, XtendedLife
  • Preparative column, XtendedLife
  • Carbon Load:
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • 11%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Max. Pressure:
  • 400 bar (5800 psi)
  • 152 bar (2200 psi)
  • 400 bar (5800 psi)
  • 400 bar (5800 psi)
  • 140 bar (2000 psi)
  • 240 bar (3500 psi)
  • 295 bar (4300 psi)
  • 140 bar (2000 psi)
  • 152 bar (2200 psi)
  • 400 bar (5800 psi)
  • 240 bar (3500 psi)
  • 240 bar (3500 psi)
  • 400 bar (5800 psi)
  • 400 bar (5800 psi)
  • 152 bar (2200 psi)
  • 240 bar (3500 psi)
  • 400 bar (5800 psi)
  • 400 bar (5800 psi)
  • 295 bar (4300 psi)
  • 400 bar (5800 psi)
  • 152 bar (2200 psi)
  • 240 bar (3500 psi)
  • 400 bar (5800 psi)
  • 152 bar (2200 psi)
  • 400 bar (5800 psi)
  • 295 bar (4300 psi)
  • 140 bar (2000 psi)
  • 295 bar (4300 psi)
  • 140 bar (2000 psi)
  • 400 bar (5800 psi)
  • 400 bar (5800 psi)
  • 152 bar (2200 psi)
  • 400 bar (5800 psi)
  • 295 bar (4300 psi)
  • 400 bar (5800 psi)
  • 295 bar (4300 psi)
  • 400 bar (5800 psi)
  • 295 bar (4300 psi)
  • 295 bar (4300 psi)
  • 295 bar (4300 psi)
  • 295 bar (4300 psi)
  • 295 bar (4300 psi)
  • 140 bar (2000 psi)
  • 152 bar (2200 psi)
  • 295 bar (4300 psi)
  • 152 bar (2200 psi)
  • 295 bar (4300 psi)
  • 295 bar (4300 psi)
  • 295 bar (4300 psi)
  • 140 bar (2000 psi)
  • 295 bar (4300 psi)
  • 240 bar (3500 psi)
  • 240 bar (3500 psi)
  • 152 bar (2200 psi)
  • 140 bar (2000 psi)
  • 152 bar (2200 psi)
  • 295 bar (4300 psi)
  • 140 bar (2000 psi)
  • 152 bar (2200 psi)
  • 295 bar (4300 psi)
  • 152 bar (2200 psi)
  • 240 bar (3500 psi)
  • pH Range:
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • Surface Area:
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • Temperature Limit:
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • USP Code:
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
Specifications