Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
D-(-)-Luciferin

D-(-)-Luciferin

500 mg, Plastic bottle
Catalog # 77510-302
Supplier:   RPI
Supplier #:  L37000-0.5
D-(-)-Luciferin
Catalog # 77510-302
Supplier:  RPI
CAS Number:  2591-17-5
D-(-)-Luciferin
D-(-)-Luciferin

500 mg, Plastic bottle

CAS Number:  2591-17-5
Price
$646.76
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Used with firefly luciferase in bioluminescence for the determination of ATP.

Caution: Research or further manufacturing use only, not for food or drug use.

Synonyms:  D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
  • CAS Number:2591-17-5
  • Formula:C₁₁H₈N₂O₃S₂
  • MDL Number:MFCD00042929
  • Molecular Weight:280.3228 g/mol
  • Melting Pt:201 °C
  • Storage Temperature:Freezer
  • EINECS:219-981-3
Specifications

Specification Test Results

  • Appearance:
    White to yellow powder
  • Biological Activity (Basic, ABS ratio of 385/285 nm):
    ≥2.5
  • Biological Activity (Neutral, ABS ratio of 327/268 nm):
    ≥2.3
  • Identification (FTIR):
    Conforms to structure
  • Purity (HPLC):
    ≥98%
  • Chiral Purity:
    ≥97.0%

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.