ContactUsLinkComponent
Monoject™ Allergy Syringes with Permanently Attached Needles, in Trays, Covidien
Monoject™ Allergy Syringes with Permanently Attached Needles, in Trays, Covidien
Catalog # 82001-194
Supplier:  Cardinal Health
undefined
Monoject™ Allergy Syringes with Permanently Attached Needles, in Trays, Covidien
Catalog # 82001-194
Supplier:  Cardinal Health
Supplier Number:  8881500501
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Color:
    Brown
  • Cat. No.:
    82001-194
  • Needle length:
    1.3 cm (1/2")
  • Supplier no.:
    8881500501
Specifications
About this item
Each tray contains 25 sterile, 28-gauge allergy syringes with an ultra-comfort bevel for injection
The syringes feature a flat plunger tip and graduations at 0.01cc. Latex free.