PIPES Sodium Salt
: RPI
- pKa (25 °C): 6.8
- Solubility (250 mg/ml, 0.1 N NaOH): Clear colorless
- Infrared Spectrum: Conforms to structure
- Water (KF): ≤2 %
- UV Absorbance 290 nm (C= 20% w/w in 0.1 N NaoH): ≤0.1
- Heavy metals (as Pb): ≤5 ppm
- Titration by H₂SO₄ (anhydrous): ≥99.0 %
- Country of origin: China
: Research or further manufacturing use only, not for food or drug use.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- CAS No.:10010-67-0
- Molecular Weight:324.35
- Storage Temperature:RT
- pH:6.5 - 7.5
- Molecular Formula:C₈H₁₇N₂NaO₆S₂
- Appearance:White to off white powder