PBS, 200 ml tablets refer to pre-weighed tablets used to prepare a specific volume of PBS (Phosphate Buffered Saline) solution. The buffer tablets are commonly used buffer solution in biological and biochemical research. PBS serves as a versatile buffer for various applications, including cell culture, immunohistochemistry, protein and nucleic acid research, and diagnostic assays. It helps maintain a stable pH and osmolarity, providing optimal conditions for biological reactions and preserving the integrity of cells and biomolecules.
- Solubility: Water
- Total weight: 1814.5 - 2005.5 mg per tablet
- Dissolution time: ≤600 seconds, Ø: ⁵/₈"
The PBS, 200 ml tablets are designed to simplify the preparation process of PBS solutions. Each tablet contains pre-determined amounts of sodium chloride, sodium phosphate dibasic, and potassium phosphate monobasic. By using these tablets, researchers can save time and ensure accuracy by eliminating the need to weigh and measure individual components.
The tablets dissolve rapidly, releasing the components and instantly forming a ready-to-use PBS solution with the desired volume. The convenience of pre-weighed tablets simplifies laboratory workflows, ensuring consistent buffer quality and reducing the potential for error in solution preparation.
: Research or further manufacturing use only, not for food or drug use.
- Pack type:100 tablets
- Color:Colorless
- Description:Phosphate buffered saline
- Size:200 ml
- pH:7.20 - 7.60
- Format:Round Tablet
- Cat. No.:77510-394
- Form:Solution
- Appearance:White round tablets
- CAS No.:7647-14-5<br />7447-40-7
- Storage Temperature:RT
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



