About this item
PBS 10X solution, is a widely used reagent in life science research. It is a buffered saline solution that contains phosphate ions, which help maintain a stable pH level in biological and biochemical experiments. PBS 10X solution is commonly used in various applications such as cell culture, immunohistochemistry, immunocytochemistry, and molecular biology techniques. The primary function of PBS 10X solution is to provide a stable environment for biological samples by maintaining a physiological pH and ionic strength.
- Solubility: Water
The phosphate buffer system present in PBS helps to resist changes in pH caused by the addition of acids or bases, making it an essential tool for experimental procedures where pH control is crucial. PBS 10X solution is prepared as a concentrated solution, typically at ten times the final desired concentration. This concentrated form allows researchers to easily dilute it to the required working concentration for their specific experiments. It is available commercially from various suppliers and comes in different formats, such as liquid or powder form.
The versatility and reliability of PBS 10X solution have made it a standard reagent in many laboratory protocols. Its use in cell biology and molecular biology research ensures consistent and reproducible results. PBS 10X solution is a common choice for washing and resuspending cells, tissues, or protein samples, as it provides the necessary osmolarity and pH stability required for these procedures.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:7732-18-5
7647-14-5
7447-40-7
7558-79-4
7778-77-0 - Storage Temperature:RT
- Appearance:Clear and colorless solution
Specifications
Related Information
Research Products International Biological Buffers RPI Biochemicals