Orders LinkContactUsLinkComponent
Vials, with Caps, External Thread
Supplier:   NEST SCIENTIFIC
Vials, with Caps, External Thread
Supplier:  NEST SCIENTIFIC
Certificates
Vials, with Caps, External Thread
2 Options Available Below
Product Details & Documents
This style of vials use with existing tube racks and automated filling equipment.
  • 1.5 ml conical vials
  • Natural color caps
  • External thread
  • Sealing ring
  • Hinged cap with sealing ring
  • Sterile
Product Family Options
With sealing ring
Product Information
  • Capacity
    Sterility
    Packaging
    Availability
    Price
  • 77508-496
    Conical vial, external thread, with sealing ring
    Conical vial, external thread, with sealing ring
    1.5 ml
    Sterile
    50/pack, 2000/case
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $433.66
    $433.66
    Case of 2,000
       $433.66
    Supplier Number: 634102N
Hinged cap with sealing ring
Product Information
  • Capacity
    Sterility
    Packaging
    Availability
    Price
  • 77508-498
    Conical vial, external thread, hinged cap with sealing ring
    Conical vial, external thread, hinged cap with sealing ring
    1.5 ml
    Sterile
    50/pack, 2000/case
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $573.34
    $573.34
    Case of 2,000
       $573.34
    Supplier Number: 634112N