Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Disodium hydrogen phosphate ≥99%

Irritant
Supplier:   Ambeed
Disodium hydrogen phosphate ≥99%
Supplier:  Ambeed
Certificates
Disodium hydrogen phosphate ≥99%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

 : for Research Use Only

Synonyms:  sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic
  • CAS Number:7558-79-4
  • Formula:Na₂HPO₄
  • MDL Number:MFCD00003496
  • Molecular Weight:141.96 g/mol
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $10.00
    $10.00
    Each(1/Itm/Bx)
       $10.00

    Supplier Number: A591134-100G
  • Glass bottle
    500 g
    Discontinued
    Supplier Number: A591134-500G
  • Glass bottle
    1000 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $23.00
    $23.00
    Each(1/Itm/Bx)
       $23.00

    Supplier Number: A591134-1000G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.